missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactic Acid 85.0+%, TCI America™

Catalog No. L022625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L022625G 25 g
L0226500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L022625G Supplier TCI America Supplier No. L022625G

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Melting Point 17°C
Color Colorless
Boiling Point 122°C
Quantity 25 g
UN Number 3265
Formula Weight 90.08
Percent Purity ≥85.0% (T)
Physical Form Liquid
Chemical Name or Material DL-Lactic Acid
EINECSNumber (2)-1369
RTECSNumber OD2800000
TSCA Yes