missing translation for 'onlineSavingsMsg'
Learn More

DL-Homoserine 98.0+%, TCI America™

Catalog No. A03191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03191G 1 g
A03195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03191G Supplier TCI America Supplier No. A03191G

Chemical Identifiers

CAS 1927-25-9
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002618,MFCD00077786
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
PubChem CID 779
ChEBI CHEBI:30653
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
SMILES N[C@@H](CCO)C(O)=O

Specifications

Melting Point 187°C
Color White
Quantity 1 g
Formula Weight 119.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Homoserine
EINECSNumber (2)-2759
TSCA No