missing translation for 'onlineSavingsMsg'
Learn More

DL-Homoserine, 98%

CAS: 1927-25-9 | C4H9NO3 | 119.12 g/mol

$116.34 - $420.14

Chemical Identifiers

CAS 1927-25-9
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002618,MFCD00077786
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
PubChem CID 779
ChEBI CHEBI:30653
IUPAC Name 2-amino-4-hydroxybutanoic acid
SMILES N[C@@H](CCO)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAH6311803
View Documents
Thermo Scientific Chemicals
H6311803
1 g
Each for $116.34
Only null left
 
AAH6311806
View Documents
Thermo Scientific Chemicals
H6311806
5 g
Each for $420.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1927-25-9
119.12
UKAUYVFTDYCKQA-VKHMYHEASA-N
779
2-amino-4-hydroxybutanoic acid
C4H9NO3
MFCD00002618,MFCD00077786
dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
CHEBI:30653
N[C@@H](CCO)C(O)=O

Specifications

188°C to 189°C
C4H9NO3
1721682
dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
N[C@@H](CCO)C(O)=O
1 g
779
119.12
Solid
1927-25-9
MFCD00002618,MFCD00077786
14,4739
UKAUYVFTDYCKQA-VKHMYHEASA-N
2-amino-4-hydroxybutanoic acid
119.12
CHEBI:30653
98%
DL-Homoserine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-661-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only