missing translation for 'onlineSavingsMsg'
Learn More

DL-Homoserine, 98%

Catalog No. AAH6311806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6311806 5 g
AAH6311803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6311806 Supplier Thermo Scientific Chemicals Supplier No. H6311806

CAS: 1927-25-9 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1927-25-9
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00002618,MFCD00077786
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym dl-homoserine, homoserine, h-dl-hoser-oh, d,l-homoserine, h-d-homoser-oh, butyric acid, 2-amino-4-hydroxy-, dl, 2-amino-4-hydroxy-butanoic acid, dl-2-amino-4-hydroxybutyric acid, d-homoserin, h-dl-hse-oh
PubChem CID 779
ChEBI CHEBI:30653
IUPAC Name 2-amino-4-hydroxybutanoic acid
SMILES N[C@@H](CCO)C(O)=O

Specifications

Melting Point 188°C to 189°C
Beilstein 1721682
Merck Index 14,4739
Quantity 5 g
Formula Weight 119.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material DL-Homoserine
EINECSNumber 217-661-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only