missing translation for 'onlineSavingsMsg'
Learn More

DL-Homocysteine thiolactone hydrochloride, 99%

Catalog No. AAL0907722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0907722 100 g
AAL0907714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0907722 Supplier Thermo Scientific Chemicals Supplier No. L0907722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6038-19-3 | C4H8ClNOS | 153.624 g/mol

DL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Homocysteine thiolactone hydrochloride is used as an intermediate of erdosteine and citiolone.

Solubility
Soluble in water (740g/L). Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 6038-19-3
Molecular Formula C4H8ClNOS
Molecular Weight (g/mol) 153.624
MDL Number MFCD00012724
InChI Key ZSEGSUBKDDEALH-UHFFFAOYSA-N
Synonym dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
PubChem CID 110753
IUPAC Name 3-aminothiolan-2-one;hydrochloride
SMILES C1CSC(=O)C1N.Cl

Specifications

Melting Point ∽204°C (decomposition)
Quantity 100 g
Beilstein 3562103
Merck Index 14,4734
Solubility Information Soluble in water (740g/L). Soluble in dimethyl sulfoxide and methanol.
Formula Weight 153.63
Percent Purity 99%
Chemical Name or Material DL-Homocysteine thiolactone hydrochloride
EINECSNumber 227-923-3
RTECSNumber XN1928400
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only