missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Histidine monohydrochloride monohydrate, 99%

Catalog No. p-7023093
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1616506 5 g
AAA1616514 25 g
AAA1616522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1616506 Supplier Thermo Scientific Chemicals Supplier No. A1616506
Only null left

CAS: 123333-71-1 | C6H12ClN3O3 | 209.63 g/mol

DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Histidine monohydrochloride monohydrate is the mixed isomer of D- and L-histidine. It may be used to evaluate the efficiency of amino acid chiral ligand exchange media and systems and in running buffers for capillary electrophoresis. It is also an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 123333-71-1
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151028
InChI Key CMXXUDSWGMGYLZ-UHFFFAOYSA-N
Synonym dl-histidine; hypochlorous acid
PubChem CID 44828786
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride;hydrate
SMILES [H+].C1=C(NC=N1)CC(C(=O)O)N.O.[Cl-]

Specifications

Melting Point ∼250°C (decomposition)
Solubility Information Insoluble in water.
Quantity 5 g
Formula Weight 209.63 (191.62 Anhydrous)
Percent Purity 99%
Chemical Name or Material DL-Histidine monohydrochloride monohydrate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only