missing translation for 'onlineSavingsMsg'
Learn More

DL-Histidine hydrochloride, monohydrate, 98%

Catalog No. AC411720250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411720250 25 g Glass Bottle
AC411721000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411720250 Supplier Thermo Scientific Chemicals Supplier No. 411720250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 123333-71-1 | C6H10ClN3O2 | 191.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123333-71-1
Molecular Formula C6H10ClN3O2
Molecular Weight (g/mol) 191.62
MDL Number MFCD00064555
InChI Key QZNNVYOVQUKYSC-JEDNCBNOSA-N
Synonym hydrogen dl-histidine chloride, 2-amino-3-1h-imidazol-5-yl propanoic acid; hydron; chloride, 2-azanyl-3-1h-imidazol-5-yl propanoic acid; hydron; chloride
PubChem CID 53395185
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydron;chloride
SMILES [H+].[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O

Specifications

Melting Point 233.0°C
Arsenic (As) 1ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 1% max.
Assay Percent Range 98%
Other Amino Acids (TLC) none
Packaging Glass Bottle
Sulfate 300ppm max.
Transmittance 96% min. (c=3, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Quantity 25 g
Formula Weight 209.64
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-Histidine hydrochloride, monohydrate, 98%
Show More Show Less
EINECSNumber 229-266-8
TSCA TSCA

RUO – Research Use Only