missing translation for 'onlineSavingsMsg'
Learn More

DL-Glyceric Acid (20% in Water, ca. 2mol/L), TCI America™

Catalog No. D060225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D060225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D060225G Supplier TCI America Supplier No. D060225G
Only null left

Chemical Identifiers

CAS 473-81-4
Molecular Formula C3H6O4
Molecular Weight (g/mol) 106.077
MDL Number MFCD00065927
InChI Key RBNPOMFGQQGHHO-UHFFFAOYSA-N
Synonym DL-2,3-Dihydroxypropionic Acid
PubChem CID 752
ChEBI CHEBI:33508
IUPAC Name 2,3-dihydroxypropanoic acid
SMILES C(C(C(=O)O)O)O

Specifications

Quantity 25 g
Formula Weight 106.08
Physical Form Liquid
Chemical Name or Material DL-Glyceric Acid (20% in Water, ca. 2mol/L)