missing translation for 'onlineSavingsMsg'
Learn More

DL-Glyceraldehyde Diethyl Acetal 95.0+%, TCI America™

Catalog No. g02165g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02165G 5 g
G021625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02165G Supplier TCI America Supplier No. G02165G
Only null left

Chemical Identifiers

CAS 10487-05-5
Molecular Formula C7H16O4
Molecular Weight (g/mol) 164.201
MDL Number MFCD00059599
InChI Key BPNZBKXZVJVEJD-UHFFFAOYSA-N
PubChem CID 542749
IUPAC Name 3,3-diethoxypropane-1,2-diol
SMILES CCOC(C(CO)O)OCC

Specifications

Color Colorless
Boiling Point 136°C
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-Glyceraldehyde Diethyl Acetal