missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Glutamine, 98%
CAS: 6899-04-3 | C5H10N2O3 | 146.146 g/mol
Supplier: Thermo Scientific Chemicals H6291503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 177°C | |
| 6899-04-3 | |
| MFCD00065103 | |
| dl-glutamine, glutamin, 2-amino-4-carbamoylbutanoic acid, dl glutamine, glutamine dl-form, glutamine, dl, l-2-aminoglutaramic acid, 2,5-bis azanyl-5-oxidanylidene-pentanoic acid, gamma-glutamine, .gamma.-glutamine | |
| C(CC(=O)N)C(C(=O)O)N | |
| 1 g | |
| 738 | |
| 146.14 |
| DL-Glutamine | |
| C5H10N2O3 | |
| 14,4471 | |
| ZDXPYRJPNDTMRX-UHFFFAOYSA-N | |
| 2,5-diamino-5-oxopentanoic acid | |
| 146.146 | |
| CHEBI:28300 | |
| 98% |
Chemical Identifiers
| 6899-04-3 | |
| 146.146 | |
| ZDXPYRJPNDTMRX-UHFFFAOYSA-N | |
| 738 | |
| 2,5-diamino-5-oxopentanoic acid |
| C5H10N2O3 | |
| MFCD00065103 | |
| dl-glutamine, glutamin, 2-amino-4-carbamoylbutanoic acid, dl glutamine, glutamine dl-form, glutamine, dl, l-2-aminoglutaramic acid, 2,5-bis azanyl-5-oxidanylidene-pentanoic acid, gamma-glutamine, .gamma.-glutamine | |
| CHEBI:28300 | |
| C(CC(=O)N)C(C(=O)O)N |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only