missing translation for 'onlineSavingsMsg'
Learn More

DL-Glutamic Acid 92.0+%, TCI America™

Catalog No. G005825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G005825G 25 g
G0058500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G005825G Supplier TCI America Supplier No. G005825G
Only null left

Chemical Identifiers

CAS 617-65-2
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00063113
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym dl-glutamic acid, glutamic acid, glutamic acid, dl, glutaminsaeure, glutamic acid dl-form, dl-glutamicacid, dl-glu, +--glutamic acid, 2-aminoglutaric acid, glutamate
PubChem CID 611
ChEBI CHEBI:18237
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Melting Point 194°C
Color White
Quantity 25 g
Formula Weight 147.13
Percent Purity ≥92.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material DL-Glutamic Acid
EINECSNumber (9)-1573
RTECSNumber LZ9648000
TSCA Yes