missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DL-Glutamic acid monohydrate, 98+%
Chemical Identifiers
| CAS | 19285-83-7 |
|---|---|
| Molecular Formula | C5H9NO4·H2O |
| Molecular Weight (g/mol) | 165.15 |
| MDL Number | MFCD00150703 |
| InChI Key | OZDAOHVKBFBBMZ-UHFFFAOYSA-N |
| Synonym | dl-glutamic acid monohydrate, a-aminoglutarate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, glutaminsaeure hydrate, +/--2-aminoglutaric acid, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-aminopentanedioic acid, oxamethane, hydrate, hydrate |
| PubChem CID | 16219385 |
| IUPAC Name | 2-aminopentanedioic acid;hydrate |
| SMILES | C(CC(=O)O)C(C(=O)O)N.O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC119930250
|
Thermo Scientific™
119930250 |
25g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC119931000
|
Thermo Scientific™
119931000 |
100g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC119930010
|
Thermo Scientific™
119930010 |
1kg | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 19285-83-7 | |
| 165.15 | |
| OZDAOHVKBFBBMZ-UHFFFAOYSA-N | |
| 16219385 | |
| C(CC(=O)O)C(C(=O)O)N.O |
| C5H9NO4·H2O | |
| MFCD00150703 | |
| dl-glutamic acid monohydrate, a-aminoglutarate, 2-aminopentanedioic acid hydrate, dl-glutamic acid hydrate, glutaminsaeure hydrate, +/--2-aminoglutaric acid, +/--2-aminoglutaric acid, dl-2-aminopentanedioic acid, 2-aminopentanedioic acid, oxamethane, hydrate, hydrate | |
| 2-aminopentanedioic acid;hydrate |
Specifications
| 19285-83-7 | |
| 98+% | |
| C5H9NO4·H2O | |
| MFCD00150703 | |
| dl-glutamic acid monohydrate, a-aminoglutarate, 2-aminopentanedioic acid hydrate, h-dl-glu-oh.h2o, glutaminsaeure hydrate, +/--2-aminoglutaric acid, +/--2-aminoglutaric acid, 2-azanylpentanedioic acid hydrate, 2-azanylpentanedioic acid hydrate, hydrate, hydrate | |
| OZDAOHVKBFBBMZ-UHFFFAOYSA-N | |
| 2-aminopentanedioic acid;hydrate | |
| 16219385 | |
| 98+% |
| Authentic | |
| Plastic bottle | |
| HO2CCH2CH2CH(NH2)CO2H·H2O | |
| 25g | |
| Solubility in water: 20.54g/L (25°C). Other solubilities: sparingly soluble in alcohol and ether | |
| C(CC(=O)O)C(C(=O)O)N.O | |
| 165.15 | |
| 165.15 | |
| DL-Glutamic acid monohydrate, 99% |
RUO â Research Use Only