missing translation for 'onlineSavingsMsg'
Learn More

DL-Glutamic acid, 95%

CAS: 617-65-2 | C5H9NO4 | 147.13 g/mol

$105.75 - $856.18

Chemical Identifiers

CAS 617-65-2
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00063113
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym dl-glutamic acid, glutamic acid, glutamic acid, dl, glutaminsaeure, glutamic acid dl-form, dl-glutamicacid, dl-glu, +--glutamic acid, 2-aminoglutaric acid, glutamate
PubChem CID 611
ChEBI CHEBI:18237
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6307514
View Documents
Thermo Scientific Chemicals
H6307514
25 g
Each for $105.75
Only null left
 
AAH6307522
View Documents
Thermo Scientific Chemicals
H6307522
100 g
Each for $247.14
Only null left
 
AAH6307536
View Documents
Thermo Scientific Chemicals
H6307536
500 g
Each for $856.18
Only null left
 
Description

Description

Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Its salt form (glutamate) is an important neurotransmitter that plays a key role in long-term potentiation and is important for learning and memory. Glutamic Acid is also a key molecule in cellular metabolism. DL-Glutamic acid is used ito study its mechanisms of crystal formation and self-assembly on surfaces and copolymer development.

Solubility
Soluble in water (miscible), HCl, ether (sparingly), ethanol (sparingly), and petroleum ether (sparingly).

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Chemical Identifiers

617-65-2
147.13
WHUUTDBJXJRKMK-UHFFFAOYNA-N
611
2-aminopentanedioic acid
C5H9NO4
MFCD00063113
dl-glutamic acid, glutamic acid, glutamic acid, dl, glutaminsaeure, glutamic acid dl-form, dl-glutamicacid, dl-glu, +--glutamic acid, 2-aminoglutaric acid, glutamate
CHEBI:18237
NC(CCC(O)=O)C(O)=O

Specifications

617-65-2
MFCD00063113
14,4469
Soluble in water (miscible),HCl,ether (sparingly),ethanol (sparingly),and petroleum ether (sparingly).
NC(CCC(O)=O)C(O)=O
25 g
611
147.13
DL-Glutamic acid
C5H9NO4
1723799
dl-glutamic acid, glutamic acid, glutamic acid, dl, glutaminsaeure, glutamic acid dl-form, dl-glutamicacid, dl-glu, +--glutamic acid, 2-aminoglutaric acid, glutamate
WHUUTDBJXJRKMK-UHFFFAOYNA-N
2-aminopentanedioic acid
147.13
CHEBI:18237
95%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-522-2

RTECSNumber : LZ9648000

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only