missing translation for 'onlineSavingsMsg'
Learn More

DL-Ethyl 2-bromopropionate, 99%

Catalog No. p-127042
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118141000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC118141000 Supplier Thermo Scientific Chemicals Supplier No. 118141000

CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 535-11-5
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000144
InChI Key ARFLASKVLJTEJD-UHFFFAOYNA-N
Synonym ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate
PubChem CID 79040
IUPAC Name ethyl 2-bromopropanoate
SMILES CCOC(=O)C(C)Br

Specifications

Color Colorless to Yellow
Density 1.3940g/mL
Boiling Point 156°C to 160°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.447
Linear Formula CH3CH(Br)CO2C2H5
Quantity 100 g
Beilstein 02,255
Specific Gravity 1.394
Merck Index 15,3827
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 181.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-Ethyl 2-bromopropionate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 208-609-5
RTECSNumber UA2451730
TSCA TSCA

RUO – Research Use Only