missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DL-Dithiothreitol

Catalog No. 315410G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 315410G Supplier Tocris Bioscience™ Supplier No. 3154/10G
Only null left

Maintains SH groups in the reduced state

Reagent for maintaining SH groups in the reduced state.

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2

Specifications

Quantity 10 g
Formula Weight 154.25
Purity >98%
Molecular Weight (g/mol) 154.24
Chemical Name or Material DL-Dithiothreitol
Recommended Storage Store at +4°C