missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol 98.0+%, TCI America™

Catalog No. D36475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36475G 5 g
D36471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36475G Supplier TCI America Supplier No. D36475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Melting Point 43°C
Color White
Boiling Point 130°C
Quantity 5 g
Formula Weight 154.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Dithiothreitol [for Electrophoresis]
RTECSNumber XO8576500
TSCA Yes
Recommended Storage Refrigerator