missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol, 99.5%, MP Biomedicals™

Catalog No. p-4405546
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19482101 1 g
ICN19482105 5 g
ICN19482110 10 g
ICN19482125 25 g
ICN19482150 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19482101 Supplier MP Biomedicals Inc Supplier No. 0219482101
Only null left

Chemical Identifiers

CAS 27565-41-9
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877,MFCD00064305
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
Synonym Cleland's reagent, DTT
IUPAC Name 1,4-disulfanylbutane-2,3-diol
SMILES OC(CS)C(O)CS

Specifications

Color White
Assay Percent Range >99%
Quantity 1 g
Formula Weight 154.2
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material DL-Dithiothreitol
EINECSNumber 248-531-9
Recommended Storage Store at + 4°C