missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Dithiothreitol, ≥99.0%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210059710
Specifications
| DL-Dithiothreitol | |
| White | |
| C4H10O2S2 | |
| 10 g | |
| VHJLVAABSRFDPM-UHFFFAOYNA-N | |
| 1,4-disulfanylbutane-2,3-diol | |
| 154.25 |
| 27565-41-9 | |
| ≥99.0% | |
| MFCD00004877,MFCD00064305 | |
| DTT, 1, 4-Dithio-DL-threitol, Cleland Reagent | |
| OC(CS)C(O)CS | |
| 154.24 | |
| Powder |
Chemical Identifiers
| 27565-41-9 | |
| 154.24 | |
| VHJLVAABSRFDPM-UHFFFAOYNA-N | |
| 1,4-disulfanylbutane-2,3-diol |
| C4H10O2S2 | |
| MFCD00004877,MFCD00064305 | |
| DTT, 1, 4-Dithio-DL-threitol, Cleland Reagent | |
| OC(CS)C(O)CS |
Safety and Handling
EINECSNumber : 248-531-9