missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol, ≥99.0%, MP Biomedicals™

Catalog No. ICN10059701
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10059701 1 g
ICN10059705 5 g
ICN10059710 10 g
ICN10059725 25 g
ICN10059750 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10059701 Supplier MP Biomedicals Inc Supplier No. 0210059701

Chemical Identifiers

CAS 27565-41-9
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877,MFCD00064305
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
Synonym DTT, 1, 4-Dithio-DL-threitol, Cleland Reagent
IUPAC Name 1,4-disulfanylbutane-2,3-diol
SMILES OC(CS)C(O)CS

Specifications

Color White
Assay Percent Range ≥99.0%
Quantity 1 g
Formula Weight 154.25
Physical Form Powder
Chemical Name or Material DL-Dithiothreitol
EINECSNumber 248-531-9