missing translation for 'onlineSavingsMsg'
Learn More

DL-Carnitine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124197
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
C005025G 25 g
C0050500G 500 g
2 options

Catalog No. C005025G

Supplier: TCI America C005025G

Only null left

Chemical Identifiers

CAS 461-05-2
Molecular Formula C7H17ClNO3+
Molecular Weight (g/mol) 198.667
MDL Number MFCD00011904
InChI Key JXXCENBLGFBQJM-UHFFFAOYSA-O
Synonym dl-carnitine hydrochloride, 3-carboxy-2-hydroxypropyl-trimethylammonium hydrochloride, 2,4-bis oxidanyl-4-oxidanylidene-butyl-trimethyl-azanium hydrochloride
PubChem CID 24206429
IUPAC Name (3-carboxy-2-hydroxypropyl)-trimethylazanium;hydrochloride
SMILES C[N+](C)(C)CC(CC(=O)O)O.Cl

Specifications

Melting Point 198°C
Color White
Quantity 25 g
Formula Weight 197.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Carnitine Hydrochloride
EINECSNumber (9)-0253&(9)-1634
RTECSNumber BP2979140
TSCA Yes