missing translation for 'onlineSavingsMsg'
Learn More

DL-Carnitine hydrochloride, 99%

Catalog No. AC108450250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108450250 25 g Plastic bottle
AC108451000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108450250 Supplier Thermo Scientific Chemicals Supplier No. 108450250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 461-05-2 | C7H15NO3·HCl | 197.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-05-2
Molecular Formula C7H15NO3·HCl
Molecular Weight (g/mol) 197.66
MDL Number MFCD00011904
InChI Key JXXCENBLGFBQJM-UHFFFAOYSA-O
Synonym dl-carnitine hydrochloride, 3-carboxy-2-hydroxypropyl-trimethylammonium hydrochloride, 2,4-bis oxidanyl-4-oxidanylidene-butyl-trimethyl-azanium hydrochloride
PubChem CID 24206429
IUPAC Name (3-carboxy-2-hydroxypropyl)-trimethylazanium;hydrochloride
SMILES C[N+](C)(C)CC(CC(=O)O)O.Cl

Specifications

Melting Point 190°C to 205°C
Color White
pH 1.8 to 2.5 (10% soln.)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula HO2CCH2CH(OH)CH2N(CH3)3Cl
Quantity 25 g
Merck Index 15, 1849
Specific Rotation −1° to +1° (20°C, 589nm) (c=1, H2O)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot ethanol,slightly soluble in cold ethanol,practically insoluble in acetone and ether
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 197.66
Percent Purity 99%
Physical Form Crystals, Powder, Needles or Chunks
Chemical Name or Material DL-Carnitine hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-309-1
RTECSNumber BP2979140
TSCA TSCA

RUO – Research Use Only