missing translation for 'onlineSavingsMsg'
Learn More

DL-Camphoroquinone, 99%

CAS: 10373-78-1 | C10H14O2 | 166.22 g/mol

Supplier:  Thermo Scientific Chemicals 154570100

Quantity 10 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC154570100


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10373-78-1
166.22
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
25208
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
C10H14O2
MFCD00064160
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
CHEBI:34607
CC1(C2CCC1(C(=O)C2=O)C)C

Specifications

DL-Camphoroquinone
198.0°C to 202.0°C
Authentic
Glass bottle
10 g
07, 581
camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
CC1(C2CCC1(C(=O)C2=O)C)C
166.22
CHEBI:34607
99%
10373-78-1
Yellow
98.5% min. (GC)
C10H14O2
MFCD00064160
15,67
VNQXSTWCDUXYEZ-UHFFFAOYSA-N
4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
25208
166.22
Fine Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-814-1

SDS
Documents

Documents

Promotions

Promotions