missing translation for 'onlineSavingsMsg'
Learn More

DL-beta-Proline hydrochloride, 95%

CAS: 953079-94-2 | C5H10ClNO2 | 151.59 g/mol

$61.26 - $156.75

Chemical Identifiers

CAS 953079-94-2
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD06801272
InChI Key OYCLYMMIZJWYJG-UHFFFAOYSA-N
Synonym pyrrolidine-3-carboxylic acid hydrochloride, beta-proline hcl, 3-pyrrolidinecarboxylic acid hcl, 3-pyrrolidinecarboxylic acid hydrochloride, pubchem16223, beta-proline hydrochloride, ksc494m2f, pyrrolidine-3-carboxylicacidhydrochloride, 3-pyrrolidine carboxylic acid hydrochloride, rs-pyrrolidine-3-carboxylic acid hydrochloride
PubChem CID 44890816
IUPAC Name pyrrolidine-3-carboxylic acid;hydrochloride
SMILES C1CNCC1C(=O)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH57365MD
View Documents
Thermo Scientific Chemicals
H57365MD
250 mg
Each for $61.26
Only null left
 
AAH5736503
View Documents
Thermo Scientific Chemicals
H5736503
1 g
Each for $156.75
Only null left
 
Description

Description

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Chemical Identifiers

953079-94-2
151.59
OYCLYMMIZJWYJG-UHFFFAOYSA-N
44890816
C1CNCC1C(=O)O.Cl
C5H10ClNO2
MFCD06801272
pyrrolidine-3-carboxylic acid hydrochloride, beta-proline hcl, 3-pyrrolidinecarboxylic acid hcl, 3-pyrrolidinecarboxylic acid hydrochloride, pubchem16223, beta-proline hydrochloride, ksc494m2f, pyrrolidine-3-carboxylicacidhydrochloride, 3-pyrrolidine carboxylic acid hydrochloride, rs-pyrrolidine-3-carboxylic acid hydrochloride
pyrrolidine-3-carboxylic acid;hydrochloride

Specifications

30°C to 38°C
C5H10ClNO2
pyrrolidine-3-carboxylic acid hydrochloride, beta-proline hcl, 3-pyrrolidinecarboxylic acid hcl, 3-pyrrolidinecarboxylic acid hydrochloride, pubchem16223, beta-proline hydrochloride, ksc494m2f, pyrrolidine-3-carboxylicacidhydrochloride, 3-pyrrolidine carboxylic acid hydrochloride, rs-pyrrolidine-3-carboxylic acid hydrochloride
C1CNCC1C(=O)O.Cl
250 mg
>110°C (230°F)
151.59
DL-beta-Proline hydrochloride
953079-94-2
MFCD06801272
OYCLYMMIZJWYJG-UHFFFAOYSA-N
pyrrolidine-3-carboxylic acid;hydrochloride
151.59
44890816
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only