missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-beta-Homoproline hydrochloride, 97%

Catalog No. p-7030866
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31749MD 250 mg
AAH3174903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31749MD Supplier Thermo Scientific Chemicals Supplier No. H31749MD
Only null left

CAS: 71985-79-0 | C6H12ClNO2 | 165.617 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71985-79-0
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.617
MDL Number MFCD11506301
InChI Key VQDACVOAOJQTPR-UHFFFAOYSA-N
Synonym pyrrolidin-2-yl-acetic acid hydrochloride, 2-pyrrolidin-2-yl acetic acid hydrochloride, pyrrolidin-2-ylacetic acid hydrochloride, 2-pyrrolidineacetic acid hydrochloride, 2-pyrrolidin-2-yl acetic acid hcl, h-d-beta-hopro-oh.hcl, 2-pyrrolidineacetic acid hcl, dl-beta-homoproline hydrochloride, 2-pyrrolidineacetic acid, hydrochloride, 2-pyrrolidinylacetic acid hydrochloride
PubChem CID 12514707
IUPAC Name 2-pyrrolidin-2-ylacetic acid;hydrochloride
SMILES C1CC(NC1)CC(=O)O.Cl

Specifications

Melting Point 171°C to 174°C
Quantity 250 mg
Formula Weight 165.62
Percent Purity 97%
Chemical Name or Material DL-beta-Homoproline hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only