missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Aspartic Acid 98.0+%, TCI America™
$45.57 - $89.23
Chemical Identifiers
| CAS | 617-45-8 |
|---|---|
| Molecular Formula | C4H7NO4 |
| Molecular Weight (g/mol) | 133.10 |
| MDL Number | MFCD00063083 |
| InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
| Synonym | dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart |
| PubChem CID | 424 |
| ChEBI | CHEBI:22660 |
| IUPAC Name | 2-aminobutanedioic acid |
| SMILES | NC(CC(O)=O)C(O)=O |
Chemical Identifiers
| C4H7NO4 | |
| MFCD00063083 | |
| dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
| CHEBI:22660 | |
| NC(CC(O)=O)C(O)=O |
Specifications
| 280°C | |
| White | |
| MFCD00063083 | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| 2-aminobutanedioic acid | |
| 424 | |
| 133.10 | |
| Crystalline Powder |
| 617-45-8 | |
| C4H7NO4 | |
| dl-aspartic acid, 2-aminosuccinic acid, dl-aminosuccinic acid, dl-asparagic acid, aspartic acid, dl, dl-asp-oh, +--aspartic acid, r,s-aspartic acid, h-dl-asp-oh, acid d,l-aspart | |
| NC(CC(O)=O)C(O)=O | |
| 133.10 | |
| CHEBI:22660 | |
| ≥98.0% (T) | |
| DL-Aspartic Acid |
Safety and Handling
EINECSNumber : (2)-1305
RTECSNumber : CI9097800
TSCA : Yes