missing translation for 'onlineSavingsMsg'
Learn More

DL-Asparagine Monohydrate 99.0+%, TCI America™

Catalog No. p-7116954
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A054125G 25 g
A0541100G 100 g
A0541500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A054125G Supplier TCI America Supplier No. A054125G
Only null left

Chemical Identifiers

CAS 3130-87-8
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00151039,MFCD00008036,MFCD00064400
InChI Key DCXYFEDJOCDNAF-UHFFFAOYNA-N
Synonym dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
PubChem CID 522362
IUPAC Name 2-amino-3-carbamoylpropanoic acid
SMILES NC(CC(N)=O)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 132.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Asparagine Monohydrate
EINECSNumber (2)-2383
TSCA Yes