missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%

CAS: 69833-18-7 | C4H8N2O3 | 132.12 g/mol

Supplier:  Thermo Scientific Chemicals B2027322

 View more versions of this product

Catalog No. AAB2027322

Only null left
Explore available promotions

Description

Description

DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

Solubility
Soluble in water, acids, and alkalis.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

∼220°C (decomposition)
69833-18-7
MFCD00151039,MFCD00008036,MFCD00064400
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
DCXYFEDJOCDNAF-UHFFFAOYNA-N
2-amino-3-carbamoylpropanoic acid
132.12
150.14 (132.12 Anhydrous)
DL-Asparagine monohydrate
C4H8N2O3
6234992
Soluble in water,acids,and alkalis.
NC(CC(N)=O)C(O)=O
100 g
522362
98%

Chemical Identifiers

69833-18-7
132.12
DCXYFEDJOCDNAF-UHFFFAOYNA-N
522362
C4H8N2O3
MFCD00151039,MFCD00008036,MFCD00064400
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
NC(CC(N)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-521-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only