missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DL-Asparagine monohydrate, 98%
Supplier: Thermo Scientific™ 167120025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| DL-Asparagine monohydrate | |
| 3130-87-8 | |
| 98% | |
| Plastic bottle | |
| MFCD00151039,MFCD00008036,MFCD00064400 | |
| 90% min. (c=10, 2N HCl, 430nm) 1cm cell | |
| dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina | |
| DCXYFEDJOCDNAF-UHFFFAOYNA-N | |
| 2,4-diamino-4-oxobutanoic acid;hydrate | |
| 132.12 | |
| 150.14 |
| 98% | |
| Authentic | |
| C4H8N2O3 | |
| H2NCOCH2CH(NH2)CO2H·H2O | |
| 04, 484 | |
| 14, 837 | |
| 10.5 to 13.5% (K.F.) | |
| NC(CC(N)=O)C(O)=O | |
| 2.5kg | |
| 522362 | |
| ≥97.5% |
Safety and Handling
EINECSNumber : 221-521-1
TSCA : TSCA
RUO â Research Use Only