missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Aminoglutethimide 98.0+%, TCI America™
$542.44 - $542.44
Chemical Identifiers
| CAS | 125-84-8 |
|---|---|
| Molecular Formula | C13H16N2O2 |
| Molecular Weight (g/mol) | 232.283 |
| MDL Number | MFCD00010122 |
| InChI Key | ROBVIMPUHSLWNV-UHFFFAOYSA-N |
| Synonym | aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum |
| PubChem CID | 2145 |
| ChEBI | CHEBI:2654 |
| IUPAC Name | 3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione |
| SMILES | CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N |
Chemical Identifiers
| 125-84-8 | |
| 232.283 | |
| ROBVIMPUHSLWNV-UHFFFAOYSA-N | |
| 2145 | |
| 3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione |
| C13H16N2O2 | |
| MFCD00010122 | |
| aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum | |
| CHEBI:2654 | |
| CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N |
Specifications
| 125-84-8 | |
| White-Yellow | |
| MFCD00010122 | |
| aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum | |
| CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N | |
| 232.283 | |
| CHEBI:2654 | |
| ≥98.0% (HPLC,T) | |
| DL-Aminoglutethimide |
| 152°C | |
| C13H16N2O2 | |
| 25 g | |
| ROBVIMPUHSLWNV-UHFFFAOYSA-N | |
| 3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione | |
| 2145 | |
| 232.28 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : MA4026950
TSCA : Yes