missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Tocopherol 96.0+%, TCI America™

Catalog No. T025125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T025125G 25 g
T0251250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T025125G Supplier TCI America Supplier No. T025125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

Melting Point 4°C
Color Brown-Yellow
Boiling Point 220°C
Quantity 25 g
Formula Weight 430.72
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-alpha-Tocopherol
EINECSNumber (9)-0864
RTECSNumber GA8746000
TSCA Yes