missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Methylbenzylamine, 99%

Catalog No. p-159806
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126391000 100 g Glass bottle
AC126395000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC126391000 Supplier Thermo Scientific Chemicals Supplier No. 126391000
Only null left

DL-alpha-Methylbenzylamine, 99%, C8H11N, CAS Number-618-36-0, 1-amino-1-phenylethane, 1-phenethylamine, 1-phenylethan-1-amine, 1-phenylethylamine, alpha-aminoethylbenzene, alpha-methylbenzenemethanamine, alpha-methylbenzylamine, alpha-phenylethylamine, a-methylbenzylamine, dl-alpha-methylbenzylamine | CAS: 618-36-0 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-36-0
MDL Number MFCD00008069
InChI Key RQEUFEKYXDPUSK-UHFFFAOYSA-N
Synonym 1-phenylethylamine, alpha-methylbenzylamine, alpha-phenylethylamine, dl-alpha-methylbenzylamine, 1-phenethylamine, alpha-aminoethylbenzene, 1-amino-1-phenylethane, 1-phenylethan-1-amine, a-methylbenzylamine, alpha-methylbenzenemethanamine
PubChem CID 7408
ChEBI CHEBI:670
IUPAC Name 1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -65.0°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 188.0°C
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H11N
Refractive Index 1.5250 to 1.5270
Linear Formula C6H5CH(CH3)NH2
Quantity 100 g
Beilstein 12, 1094
Specific Gravity 0.94
Merck Index 14, 6026
Solubility Information Solubility in water: 4.2% (20°C). Other solubilities: miscible with alcohol and ether,soluble in most organic solvents
Molecular Weight (g/mol) 121.18
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-α-Methylbenzylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 210-545-8
TSCA TSCA

RUO – Research Use Only