missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Amino-epsilon-caprolactam 98.0+%, TCI America™

$52.19 - $177.08

Chemical Identifiers

CAS 671-42-1
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00064475
InChI Key BOWUOGIPSRVRSJ-UHFFFAOYSA-N
Synonym DL-3-Aminohexahydro-2-azepinone, DL-3-Amino-2-oxohexamethyleneimine
PubChem CID 102463
ChEBI CHEBI:19471
IUPAC Name 3-aminoazepan-2-one
SMILES C1CCNC(=O)C(C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A10031G
View Documents
TCI America
A10031G
1 g N/A
Only null left
 
A10035G
View Documents
TCI America
A10035G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

671-42-1
128.175
BOWUOGIPSRVRSJ-UHFFFAOYSA-N
102463
3-aminoazepan-2-one
C6H12N2O
MFCD00064475
DL-3-Aminohexahydro-2-azepinone, DL-3-Amino-2-oxohexamethyleneimine
CHEBI:19471
C1CCNC(=O)C(C1)N

Specifications

671-42-1
White-Yellow
C6H12N2O
1 g
BOWUOGIPSRVRSJ-UHFFFAOYSA-N
3-aminoazepan-2-one
102463
128.18
Crystalline Powder
77°C
172°C
MFCD00064475
DL-3-Aminohexahydro-2-azepinone, DL-3-Amino-2-oxohexamethyleneimine
C1CCNC(=O)C(C1)N
128.175
CHEBI:19471
≥98.0% (GC)
DL-alpha-Amino-epsilon-caprolactam
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-1103

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'