missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Amino-epsilon-caprolactam 98.0+%, TCI America™

Catalog No. A10035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10035G 5 g
A10031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10035G Supplier TCI America Supplier No. A10035G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 671-42-1
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00064475
InChI Key BOWUOGIPSRVRSJ-UHFFFAOYSA-N
Synonym DL-3-Aminohexahydro-2-azepinone, DL-3-Amino-2-oxohexamethyleneimine
PubChem CID 102463
ChEBI CHEBI:19471
IUPAC Name 3-aminoazepan-2-one
SMILES C1CCNC(=O)C(C1)N

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 128.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material DL-alpha-Amino-epsilon-caprolactam
EINECSNumber (5)-1103
TSCA Yes