missing translation for 'onlineSavingsMsg'
Learn More

DL-alpha-Amino-2-thiopheneacetic acid methyl ester hydrochloride, 98%

Catalog No. p-7028066
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2519003 1 g
AAB2519006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2519003 Supplier Thermo Scientific Chemicals Supplier No. B2519003

CAS: 132289-66-8 | C7H10ClNO2S | 207.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132289-66-8
Molecular Formula C7H10ClNO2S
Molecular Weight (g/mol) 207.67
MDL Number MFCD04039779
InChI Key NZEQIXANVTXFEP-UHFFFAOYNA-N
Synonym methyl 2-amino-2-thiophen-2-yl acetate hydrochloride, dl-alpha-aminothiophene-2-acetic acid methyl ester hydrochloride
PubChem CID 24212017
SMILES Cl.COC(=O)C(N)C1=CC=CS1

Specifications

Melting Point ∼180°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
IUPAC Name methyl 2-amino-2-(thiophen-2-yl)acetate hydrochloride
Formula Weight 207.67
Percent Purity 98%
Chemical Name or Material DL-alpha-Amino-2-thiopheneacetic acid methyl ester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only