missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Alanine, 99%

Catalog No. AAA1223030
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1223030 250 g
AAA122300B 1000 g
AAA122300I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1223030 Supplier Thermo Scientific Chemicals Supplier No. A1223030

CAS: 302-72-7 | C3H7NO2 | 89.09 g/mol

DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 302-72-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064408
InChI Key QNAYBMKLOCPYGJ-UHFFFAOYNA-N
Synonym dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
PubChem CID 602
ChEBI CHEBI:16449
IUPAC Name 2-aminopropanoic acid
SMILES CC(N)C(O)=O

Specifications

Melting Point ∼292°C (decomposition)
Beilstein 635807
Merck Index 14,204
Solubility Information Soluble in water. Slightly soluble in alcohol.
Density 1.42
Quantity 250 g
Formula Weight 89.09
Percent Purity 99%
Chemical Name or Material DL-Alanine
EINECSNumber 206-126-4
RTECSNumber AY2980000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only