missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Alanine, 99%

CAS: 302-72-7 | C3H7NO2 | 89.09 g/mol

Supplier:  Thermo Scientific Chemicals A122300I

Quantity 5000 g

 View more versions of this product

Catalog No. AAA122300I


Only null left

Description

Description

DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

302-72-7
89.09
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
602
2-aminopropanoic acid
C3H7NO2
MFCD00064408
dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
CHEBI:16449
CC(N)C(O)=O

Specifications

∼292°C (decomposition)
302-72-7
MFCD00064408
14,204
Soluble in water. Slightly soluble in alcohol.
CC(N)C(O)=O
2-aminopropanoic acid
5000 g
CHEBI:16449
99%
DL-Alanine
C3H7NO2
635807
dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine
QNAYBMKLOCPYGJ-UHFFFAOYNA-N
1.42
89.09
602
89.09
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-126-4

RTECSNumber : AY2980000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions