Learn More
Thermo Scientific Chemicals DL-Alanine, 99%
CAS: 302-72-7 | C3H7NO2 | 89.09 g/mol
Supplier: Thermo Scientific Chemicals A122300B
| Quantity | 1000 g |
|---|
Description
DL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Alanine is used in mixed ligand (chelate) studies with transition metals such as Cu(II), Zn(ll) and Cd(ll). It is a functional component in the alanine cycle which regulates the level of alanine aminotransferases which can be used as a diagnostic tool to predict type II diabetes. It is used as a model reagent for development of amino acid racemic resolution techniques.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| C3H7NO2 | |
| MFCD00064408 | |
| dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine | |
| CHEBI:16449 | |
| CC(N)C(O)=O |
Specifications
| ∼292°C (decomposition) | |
| 302-72-7 | |
| MFCD00064408 | |
| 14,204 | |
| Soluble in water. Slightly soluble in alcohol. | |
| CC(N)C(O)=O | |
| 2-aminopropanoic acid | |
| 1000 g | |
| CHEBI:16449 | |
| 99% |
| DL-Alanine | |
| C3H7NO2 | |
| 635807 | |
| dl-alanine, alanine, alanine, dl, dl-alpha-alanine, d,l-alanine, dl-2-aminopropionic acid, r,s-alanine, dl-2-aminopropanoic acid, h-dl-ala-oh, +--alanine | |
| QNAYBMKLOCPYGJ-UHFFFAOYNA-N | |
| 1.42 | |
| 89.09 | |
| 602 | |
| 89.09 |
Safety and Handling
EINECSNumber : 206-126-4
RTECSNumber : AY2980000
TSCA : Yes
Recommended Storage : Ambient temperatures