missing translation for 'onlineSavingsMsg'
Learn More

DL-4-Hydroxy-3-methoxymandelic Acid 98.0+%, TCI America™

Catalog No. H02681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02681G 1 g
H02685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02681G Supplier TCI America Supplier No. H02681G

Chemical Identifiers

CAS 55-10-7
Molecular Formula C9H9O5
Molecular Weight (g/mol) 197.17
MDL Number MFCD00004235
InChI Key CGQCWMIAEPEHNQ-QMMMGPOBSA-M
Synonym vanillylmandelic acid, vanilmandelic acid, dl-4-hydroxy-3-methoxymandelic acid, 2-hydroxy-2-4-hydroxy-3-methoxyphenyl acetic acid, 4-hydroxy-3-methoxymandelic acid, vanillomandelic acid, +/--vanillylmandelic acid, vma, alpha,4-dihydroxy-3-methoxybenzeneacetic acid, benzeneacetic acid, .alpha.,4-dihydroxy-3-methoxy
PubChem CID 1245
ChEBI CHEBI:20106
IUPAC Name (2S)-2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetate
SMILES COC1=CC(=CC=C1O)[C@H](O)C([O-])=O

Specifications

Melting Point 133°C
Quantity 1 g
Formula Weight 198.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-4-Hydroxy-3-methoxymandelic Acid