missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Methylcyclopentanone, 99%

Catalog No. AC169370050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169370050 5 g Glass bottle
AC169370250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169370050 Supplier Thermo Scientific Chemicals Supplier No. 169370050
Only null left

CAS: 1757-42-2 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1757-42-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00001416
InChI Key AOKRXIIIYJGNNU-UHFFFAOYSA-N
Synonym 3-methylcyclopentanone, cyclopentanone, 3-methyl, dl-3-methylcyclopentanone, 3-methyl-1-cyclopentanone, cyclopentanone, 3-methyl-, r, 3-methylcyclopentanone, +/-, acmc-20apjx, 3-methyl cyclopentanone, 3-methyl-cyclopentanone, acmc-1ba4k
PubChem CID 15650
IUPAC Name 3-methylcyclopentan-1-one
SMILES CC1CCC(=O)C1

Specifications

Color Yellow
Density 0.9100g/mL
Boiling Point 145°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4330 to 1.435
Linear Formula CH3C5H7(=O)
Quantity 5 g
Beilstein 07, 11
Specific Gravity 0.91
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ether and acetone
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material DL-3-Methylcyclopentanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-148-9

RUO – Research Use Only