missing translation for 'onlineSavingsMsg'
Learn More

DL-3-Hydroxybutyric acid, sodium salt, 99%

Catalog No. p-3758186
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215010250 25 g Glass bottle
AC215011000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC215010250 Supplier Thermo Scientific Chemicals Supplier No. 215010250
Only null left

CAS: 150-83-4 | C4H7NaO3 | 126.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-83-4
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00016716
InChI Key NBPUSGBJDWCHKC-UHFFFAOYSA-M
Synonym sodium 3-hydroxybutyrate, sodium 3-hydroxybutanoate, dl-3-hydroxybutyric acid sodium salt, sodium beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, monosodium salt, butanoic acid,3-hydroxy-, sodium salt 1:1, dl-3-hydroxybutyric acid, sodium salt, 3-hydroxybutyric acid sodium salt, sodium 3-oxidanylbutanoate, dl-beta-hydroxybutyric acid sodium salt
PubChem CID 23676771
IUPAC Name sodium;3-hydroxybutanoate
SMILES CC(CC(=O)[O-])O.[Na+]

Specifications

Melting Point 161.0°C to 167.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2CO2Na
Quantity 25 g
Beilstein 03, 308
Solubility Information Solubility in water: freely soluble
Formula Weight 126.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-3-Hydroxybutyric acid, sodium salt
Show More Show Less
EINECSNumber 205-774-5

RUO – Research Use Only