missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-3-Hydroxybutyric Acid (contains Polymolecular esterification product) 80.0+%, TCI America™
$111.66 - $375.41
Chemical Identifiers
| CAS | 300-85-6 |
|---|---|
| Molecular Formula | C4H8O3 |
| Molecular Weight (g/mol) | 104.105 |
| MDL Number | MFCD00004546 |
| InChI Key | WHBMMWSBFZVSSR-UHFFFAOYSA-N |
| Synonym | 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy |
| PubChem CID | 441 |
| ChEBI | CHEBI:20067 |
| IUPAC Name | 3-hydroxybutanoic acid |
| SMILES | CC(CC(=O)O)O |
Chemical Identifiers
| 300-85-6 | |
| 104.105 | |
| WHBMMWSBFZVSSR-UHFFFAOYSA-N | |
| 441 | |
| 3-hydroxybutanoic acid |
| C4H8O3 | |
| MFCD00004546 | |
| 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy | |
| CHEBI:20067 | |
| CC(CC(=O)O)O |
Specifications
| 300-85-6 | |
| C4H8O3 | |
| 5 g | |
| WHBMMWSBFZVSSR-UHFFFAOYSA-N | |
| 3-hydroxybutanoic acid | |
| 441 | |
| 104.11 | |
| Liquid |
| Yellow | |
| MFCD00004546 | |
| 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy | |
| CC(CC(=O)O)O | |
| 104.105 | |
| CHEBI:20067 | |
| ≥80.0% (T) | |
| DL-3-Hydroxybutyric Acid (contains Polymolecular esterification product) |
Safety and Handling
EINECSNumber : (2)-1429
TSCA : Yes
Recommended Storage : Refrigerator