missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylpropionic Acid 98.0+%, TCI America™

Catalog No. P06995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06995G 5 g
P069925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06995G Supplier TCI America Supplier No. P06995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 492-37-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002650
InChI Key YPGCWEMNNLXISK-UHFFFAOYSA-N
Synonym 2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid
PubChem CID 10296
ChEBI CHEBI:48526
IUPAC Name 2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 267°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material DL-2-Phenylpropionic Acid
EINECSNumber (3)-1751
TSCA Yes