missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Phenylglycinol, 98%

Catalog No. p-4510695
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC374280050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC374280050 Supplier Thermo Scientific Chemicals Supplier No. 374280050

CAS: 7568-92-5 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7568-92-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00130145
InChI Key IJXJGQCXFSSHNL-UHFFFAOYNA-N
Synonym 2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
PubChem CID 92466
SMILES NC(CO)C1=CC=CC=C1

Specifications

Melting Point 53.0°C to 58.0°C
Color White to Yellow
Boiling Point 261.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5484
Linear Formula C6H5CH(NH2)CH2OH
Quantity 5 g
Beilstein 13,IV,1835
IUPAC Name 2-amino-2-phenylethan-1-ol
Formula Weight 137.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material DL-2-Phenylglycinol
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only