missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Octanol, 97%

CAS: 123-96-6 | C8H18O | 130.23 g/mol

$54.74 - $268.59

Chemical Identifiers

CAS 123-96-6
MDL Number MFCD00004591
InChI Key SJWFXCIHNDVPSH-UHFFFAOYSA-N
Synonym 2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
PubChem CID 20083
ChEBI CHEBI:37869
IUPAC Name octan-2-ol
SMILES CCCCCCC(C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129411000
SDS View Documents
Thermo Scientific Chemicals
129411000
100 mL Glass bottle
Each for $54.74
Only null left
 
AC129410010
SDS View Documents
Thermo Scientific Chemicals
129410010
1 L Glass bottle
Each for $105.79
Only null left
 
AC129410025
SDS View Documents
Thermo Scientific Chemicals
129410025
2.5 L Glass bottle
Each for $268.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-96-6
SJWFXCIHNDVPSH-UHFFFAOYSA-N
20083
octan-2-ol
MFCD00004591
2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
CHEBI:37869
CCCCCCC(C)O

Specifications

123-96-6
Colorless to Yellow
174°C to 181°C
Authentic
Glass bottle
1.4250 to 1.427
MFCD00004591
01, 419
0.81
2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
SJWFXCIHNDVPSH-UHFFFAOYSA-N
octan-2-ol
20083
130.23
Liquid
-38°C
0.8100g/mL
72°C
96% min. (GC)
C8H18O
CH3(CH2)5CH(OH)CH3
100 mL
01,109
15, 6841
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether,soluble in most organic solvents
CCCCCCC(C)O
130.23
CHEBI:37869
97%
DL-2-Octanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 204-667-

SDS
Documents

Documents

RUO – Research Use Only