missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methylbutyric Acid 97.0+%, TCI America™

Catalog No. p-7132040
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M018125ML 25 mL
M0181500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M018125ML Supplier TCI America Supplier No. M018125ML

Chemical Identifiers

CAS 116-53-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00002669
InChI Key WLAMNBDJUVNPJU-UHFFFAOYSA-N
Synonym 2-methylbutyric acid, dl-2-methylbutyric acid, butanoic acid, 2-methyl, ethylmethylacetic acid, methylbutyric acid, 2-methyl butyric acid, 2-methybutyric acid, methylethylacetic acid, active valeric acid, butyric acid, 2-methyl
PubChem CID 8314
ChEBI CHEBI:37070
IUPAC Name 2-methylbutanoic acid
SMILES CCC(C)C(=O)O

Specifications

Color Yellow
Boiling Point 174°C
Quantity 25 mL
UN Number 3265
Formula Weight 102.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material DL-2-Methylbutyric Acid
EINECSNumber (2)-0608
RTECSNumber EK7897000
TSCA Yes