missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methyl-1-butanol, 98%

CAS: 137-32-6 | C5H12O | 88.15 g/mol

Supplier:  Thermo Scientific Chemicals 158270025

 View more versions of this product

Catalog No. AC158270025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

DL-2-Methyl-1-butanol
137-32-6
Colorless to Yellow
130.0°C
Authentic
Glass bottle
1.4090 to 1.4110
MFCD00004743
01, 388
15, 6104
Solubility in water: 3.6g/100mL (30°C). Other solubilities: miscible with alcohol and ether,soluble in most organic solvents
CCC(C)CO
88.15
5.1 mPa.s (20°C)
88.15
Liquid
98%
-70.0°C
0.8100g/mL
43°C
97.5% min. (GC)
C5H12O
C2H5CH(CH3)CH2OH
2.5 L
0.81
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
QPRQEDXDYOZYLA-UHFFFAOYSA-N
2-methylbutan-1-ol
8723
CHEBI:48945
98%

Chemical Identifiers

137-32-6
88.15
QPRQEDXDYOZYLA-UHFFFAOYSA-N
8723
2-methylbutan-1-ol
C5H12O
MFCD00004743
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
CHEBI:48945
CCC(C)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

EINECSNumber : 205-289-9

RTECSNumber : EL5250000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only