missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methyl-1-butanol, 98%

Catalog No. AC158270250
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158270250 25 mL Glass bottle
AC158271000 100 mL Glass bottle
AC158270010 1 L Glass bottle
AC158270025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC158270250 Supplier Thermo Scientific Chemicals Supplier No. 158270250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 137-32-6 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137-32-6
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004743
InChI Key QPRQEDXDYOZYLA-UHFFFAOYSA-N
Synonym 2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
PubChem CID 8723
ChEBI CHEBI:48945
IUPAC Name 2-methylbutan-1-ol
SMILES CCC(C)CO

Specifications

Melting Point -70.0°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 130.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4090 to 1.4110
Linear Formula C2H5CH(CH3)CH2OH
Quantity 25 mL
Beilstein 01, 388
Specific Gravity 0.81
Merck Index 15, 6104
Solubility Information Solubility in water: 3.6g/100mL (30°C). Other solubilities: miscible with alcohol and ether,soluble in most organic solvents
Viscosity 5.1 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material DL-2-Methyl-1-butanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 205-289-9
RTECSNumber EL5250000
TSCA TSCA

RUO – Research Use Only