missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-2-Benzylserine, 97%

Catalog No. AC339780010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC339780010 1 g Glass bottle
AC339780100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC339780010 Supplier Thermo Scientific Chemicals Supplier No. 339780010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4740-47-0 | C10H13NO3 | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4740-47-0
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD01863305
InChI Key ZMNNAJIBOJDHAF-JTQLQIEISA-N
Synonym dl-2-benzylserine, 2-amino-3-hydroxy-2-benzylpropanoic acid, alpha-benzyl-dl-serine
PubChem CID 7010088
IUPAC Name (2S)-2-amino-2-benzyl-3-hydroxypropanoic acid
SMILES C1=CC=C(C=C1)CC(CO)(C(=O)O)N

Specifications

Melting Point 245°C to 263°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water
Formula Weight 195.22
Percent Purity 97%
Physical Form Powder or Flakes
Chemical Name or Material DL-2-Benzylserine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only