missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Aminobutyric acid, 99%

CAS: 2835-81-6 | C4H9NO2 | 103.12 g/mol

$86.66 - $86.66

Chemical Identifiers

CAS 2835-81-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
InChI Key QWCKQJZIFLGMSD-UHFFFAOYSA-N
Synonym dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid
PubChem CID 6657
ChEBI CHEBI:35621
IUPAC Name 2-aminobutanoic acid
SMILES CCC(C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172090250
SDS View Documents
Thermo Scientific Chemicals
172090250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2835-81-6
103.12
dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid
CHEBI:35621
CCC(C(=O)O)N
C4H9NO2
QWCKQJZIFLGMSD-UHFFFAOYSA-N
6657
2-aminobutanoic acid

Specifications

2835-81-6
White
99%
C4H9NO2
25 g
15,423
Solubility in water: 210g/L (25 c). Other solubilities: sparingly soluble in alcohol,insoluble in ether
CCC(C(=O)O)N
103.12
CHEBI:35621
99%
DL-2-Aminobutyric acid, 99+%
291.0°C to 293.0°C
Authentic
Glass bottle
C2H5CH(NH2)CO2H
04,408
dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid
QWCKQJZIFLGMSD-UHFFFAOYSA-N
2-aminobutanoic acid
6657
103.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-616-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only