missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-2-Aminobutyric acid, 99%
CAS: 2835-81-6 | C4H9NO2 | 103.121 g/mol
Supplier: Thermo Scientific Chemicals L0603522
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| DL-2-Aminobutyric acid | |
| ∽291°C (decomposition) | |
| MFCD00008093 | |
| 635889 | |
| dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid | |
| CCC(C(=O)O)N | |
| 103.121 | |
| CHEBI:35621 | |
| 99% |
| 2835-81-6 | |
| C4H9NO2 | |
| 100 g | |
| 14,428 | |
| QWCKQJZIFLGMSD-UHFFFAOYSA-N | |
| 2-aminobutanoic acid | |
| 6657 | |
| 103.12 |
Chemical Identifiers
| 2835-81-6 | |
| 103.121 | |
| QWCKQJZIFLGMSD-UHFFFAOYSA-N | |
| 6657 | |
| 2-aminobutanoic acid |
| C4H9NO2 | |
| MFCD00008093 | |
| dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid | |
| CHEBI:35621 | |
| CCC(C(=O)O)N |
Safety and Handling
EINECSNumber : 220-616-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only